missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, DILUT-IT Concentrate, 0.01M, J.T. Baker™
Click to view available options
Quantity:
For dilution to 1 L
Packaging:
LDPE ampule
Description
DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.
- Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
- Space saving: Products require only one-tenth of the space required for ready-made solutions
- Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate
Specifications
Color | Colorless |
Boiling Point | 100°C |
CAS | 1310-73-2 |
Melting Point | 0°C |
Molecular Formula | HNaO |
Concentration | 0.01mol/L |
Density | 1g/cm3 at 20°C |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
Solubility Information | Fully miscible in water |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Show More |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'