missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hexafluorophosphate, 98%, Thermo Scientific Chemicals
28.26 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong acids, bases and oxidizing agents.
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong acids, bases and oxidizing agents.
Specifications
Melting Point | >200°C |
Quantity | 5 g |
UN Number | UN3260 |
Solubility Information | Soluble in water. |
IUPAC Name | sodium hexafluoro-λ⁵-phosphanuide |
Formula Weight | 167.95 |
Percent Purity | 98% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 2.37 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'