missing translation for 'onlineSavingsMsg'
Learn More

Sodium diphenylamine-4-sulfonate, 0.005M solution in water, Thermo Scientific™

Catalog Number p-7058570
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
6152-67-6
C12H11NNaO3S
272.27
MFCD00036468
VMVSDVIYIMNZBS-UHFFFAOYSA-N
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
517002
sodium;4-anilinobenzenesulfonate
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
This item is not returnable. View return policy
6152-67-6
C12H11NNaO3S
272.27
MFCD00036468
VMVSDVIYIMNZBS-UHFFFAOYSA-N
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
517002
sodium;4-anilinobenzenesulfonate
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Redox indicator

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Odor Odorless
Quantity 100 mL
Solubility Information Fully miscible with water.
Formula Weight 271.27
Concentration or Composition (by Analyte or Components) 0.005M solution in water
Physical Form Liquid
Chemical Name or Material Sodium diphenylamine-4-sulfonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'