missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99+%, extra pure, Thermo Scientific™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7775-09-9
ClNaO3
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
516902
CHEBI:65242
sodium;chlorate
[O-]Cl(=O)=O.[Na+]
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7775-09-9
ClNaO3
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
516902
CHEBI:65242
sodium;chlorate
[O-]Cl(=O)=O.[Na+]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Oxidation reagent
Quantity 250g
Assay Percent Range 99+%
Linear Formula NaClO3
Fieser 01,1056
Merck Index 15,8733
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and glycerin,7.7g/L ethanol (20°C)
Formula Weight 106.44
Physical Form Crystals
Percent Purity 99+%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 2.49
Density 2.4900g/mL
Chemical Name or Material Sodium chlorate
Show More Show Less

RUO – Research Use Only