missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Soda Lime Glass Measuring Pipet For Partial Flow

Brand:  Duran™ 243431604

Additional Details : Weight : 0.25200kg

 View more versions of this product

Product Code. 15281949

  • 59.65 EUR / Pack of 12


Estimated Shipment
14-05-2019
Add to basket

Specifications

Specifications

±0.015mL
B
Yes
360mm
Measuring Pipette
Soda Lime Glass
DIN 12 621, ISO 835
Black
0.02mL
12Pack
2mL
Videos