missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS3 Monoclonal Antibody (OTI11C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502959
This item is not returnable.
View return policy
Description
Accumulating evidence demonstrates that cytokine receptor signaling is negatively regulated by a family of Src homology 2 domain-containing adaptor molecules Termed SOCS (Suppressor of Cytokine Signaling). To date, there are eight members of SOCS family have been recognized, they are SOCS-1, 2, 3, 4, 5, 6, 7 and CIS. Structurally the SOCS proteins are composed of an N-terminal region of variable length and amino acid composition, a central SH2 domain, and a previously unrecognized C-terminal motif that we have called the SOCS box. The SOCS proteins appear to form part of a classical negative feed back loop that regulates cytokine signal transduction via a STAT-induced transcriptional mechanism. Transcription of each of the SOCS genes occurs rapidly in vitro and in vivo in response to cytokines, and once produced, the various members of the SOCS family appear to inhibit signaling in different ways.Specifications
| SOCS3 | |
| Monoclonal | |
| Unconjugated | |
| SOCS3 | |
| ATOD4, CIS3, Cish3, SOCS-3, SSI-3, SSI3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9021 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI11C5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14543 | |
| SOCS3 | |
| Full length human recombit protein of human SOCS3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction