Learn More
Heathrow Scientific™ Paper Weigh Boats
Heathrow Scientific Paper Weigh Boats are a truly green solution to sample weighing. They are made from FSC™ certified (FSC™ mix credit) paper (Eco-IN) and when disposed of in the standard waste stream they will naturally biodegrade (Eco-OUT). Controls contents like a polystyrene weigh boat.
51.65 EUR - 52.95 EUR
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
15936495
|
Heathrow Scientific™
120195 |
White |
52.95 EUR
Pack of 250 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
15893967
|
Heathrow Scientific™
120626 |
Brown |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- An alternative to regular weighing papers as they are much easier to handle
- Made of grease proof paper that meets FDA 21 CFR 176.170 (components of paper in contact with aqueous and fatty foods) and with 2I CFR 176.180 (contact with dry foods)
- Compatible with heat applications with a temperature limitation of 210°C
Specifications
Paper | |
Square | |
Weighing Boat | |
Large |