Learn More
Thermo Scientific™ SLIPFREE Ferrules
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
16.75 EUR - 21.74 EUR
Specifications
For Use With (Equipment) | SLIPFREE Connectors for HPLC Columns |
---|---|
Type | Sealing Ferrule |
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
10140643
|
Thermo Scientific™
36024 |
Vespel |
21.74 EUR
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10404783
|
Thermo Scientific™
36023 |
PEEK |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
- Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
- Kel-F and PEEK are offered for applications in which Vespel is not suitable
Specifications
SLIPFREE Connectors for HPLC Columns | |
Sealing Ferrule |