missing translation for 'onlineSavingsMsg'
Learn More

NRG1 Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the EGF-like domain (Thr 176-Lys 246) of human NRG1 (Q02297-6) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Invitrogen 11609H01H25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15258715

  • 157.00 EUR / 25µg


Estimated Shipment
05-08-2020
Add to basket

Description

Description

Neuregulin 1 (NRG1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized Rhesus ErbB3 at 2μg/mL (100μL/well) can bind human NRG1 (isoform Beta1), the EC50 of human NRG1 (isoform Beta1) is typically 0.58μg/mL; 2. Immobilized human ErbB3 at 2μg/mL (100μL/well) can bind human NRG1 (isoform Beta1), the EC50 of human NRG1 (isoform Beta1) is typically 0.43μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Other Growth Factors
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
36.7kDa
5 x 5μg
<1 EU/μg
Neuregulin 1
IgG1 Fc Tag
Growth Factors & Receptors
Assay Standard
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, NDF, SMDF
NRG1
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.