missing translation for 'onlineSavingsMsg'
Learn More

HIST1H3A Recombinant Human Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the native human HIST1H3A (P6843) (Met 1-Ala 136)

Brand:  Invitrogen 11231HNAE50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258435

  • 645.00 EUR / 50µg


Estimated Shipment
20-08-2020
Add to basket

Description

Description

Histone H3.1 (HIST1H3A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 2mM β-Mercaptoethanol, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

E. coli
Assay Standard
Lyophilized
15.5kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Histone cluster 1, H3a
Histones
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
8350
PAGE
50μg
Full Length/Recombinant
HIST1H3A, H3/A, H3FA, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3E, HIST1H3F, HIST1H3G
HIST1H3A
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.