missing translation for 'onlineSavingsMsg'
Learn More

FLRT2 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human FLRT2 (O43155) extracellular domain (Met 1-Ser 539) fused to a C-terminal polyhistidine tag

Brand:  Invitrogen 11296H08H50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258495

  • 246.00 EUR / 50µg


Estimated Shipment
25-08-2020
Add to basket

Description

Description

Fibronectin Leucine Rich Transmembrane Protein 2 (FLRT2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Assay Standard
Lyophilized
57.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
UNQ232/PRO265, KIAA0405
FLRT2
Cell Structure/Organelle Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
23768
PAGE
50μg
C-Terminal
<1 EU/μg
Fibronectin leucine rich transmembrane protein 2
His Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.