missing translation for 'onlineSavingsMsg'
Learn More

CDH17 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human CDH17 (Q12864-1) extracellular domain (Met 1-Met 787) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11360H08H25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15258545

  • 181.00 EUR / 25µg


Estimated Shipment date
from Supplier 11-08-2020
Add to basket

Description

Description

Cadherin 17, LI cadherin (liver-intestine) (CDH17), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of MCF-7 human breast adenocarcinoma cells; when 5 x 104 cells/well are added to recombinant human Cadherin 17 coated plates (0.8μg/mL with 100μL/well), approximately >50% will adhere after 1 hour at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Intercellular Adhesion Molecule
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
87kDa
5 x 5μg
<1 EU/μg
Cadherin 17, LI cadherin (liver-intestine)
His Tag
Cell Structure/Organelle Proteins
Assay Standard
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
CDH16, FLJ26931, HPT-1, HPT1, mgC138218, mgC142024
CDH17
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.