Learn More
Simport™ Scientific Ino-Loop™ Inoculating Loops and Needles
Brand:  Simport™ Scientific L200-2A
Additional Details : Weight : 1.52000kg
 View more versions of this product
Product Code. 11768992
  • 146.00 EUR / Pack of 1000
Estimated Shipment
02-02-2021
Add to basket
Description
Description
  • Smooth and flexible
  • Needles are straightand suitable for removal of specimens of single colonies
  • Packed sterile in safe, tamperproof, zip-lock resealable bags
  • Does not require flaming
  • Eliminates risk of infection due to aerosol formation of pathogenic substances
  • Used under hoods without danger
  • Color-coded for ease of size identification
  • Total length: 22.7cm
Specifications
Specifications
Yes
Polystyrene
Inoculating Loop
Yellow
Tamperproof Ziplock resealable bag
22.7cm
Sterile
10μL
10/Pk.