missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver sulfate-Sulfuric acid solution, Volumetric solution, for COD determination according to DIN 38409, part 41, Honeywell Fluka™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
- Solutions are prepared with raw materials which meet or exceed requirements of the American Chemical Society
- Where no ACS specifications exist, chemicals of highest purity available are used
- All aqueous solutions are prepared using high purity, deionized, water which meets requirements for ASTM type 1 Reagent Grade Water
- Exacting production and quality control records are maintained providing traceability of raw materials
- All solutions are guaranteed to meet label specifications
- All solution labels meet or exceed all ISO, DOT and OSHA regulations
Specifications
Quantity | 1 L |
UN Number | UN3264 |
Chemical Name or Material | Silver sulfate-Sulfuric acid solution |
Safety and Handling
Product Identifier
- Silver sulfate-Sulfuric acid solution
- Danger
- Skin corrosion/irritation Category 1 A
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sulphuric acid, Disilver(1+) sulphate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'