missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog Number p-221911
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
Boiling Point 444.0°C
Melting Point 212.0°C
Color Green to Red
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99.4% min. (Argentometry)
Linear Formula AgNO3
Fieser 01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39
Merck Index 15, 8657
Solubility Information Solubility in water: 219 g/100mL (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone and ether
Formula Weight 169.87
Percent Purity 99.5%
Grade Analytical
Identification Passes Test
Packaging Glass bottle
Decomposition Information >444°C
Density 4.35
Chemical Name or Material Silver nitrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Silver nitrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'