missing translation for 'onlineSavingsMsg'
Learn More

Silver nanoparticles, 20nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 405nm absorption, Thermo Scientific Chemicals

Catalog Number p-7057630
Click to view available options
Quantity:
25 mL
100 mL
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

Solubility
Fully miscible.

Notes
Particles per ml: 6.2E+11
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 25 mL
Solubility Information Fully miscible.
Odor Odorless
Particle Size 20nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Silver nanoparticles, 0.02mg/mL, supplied in 2mM sodium citrate, 405nm absorption

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'