missing translation for 'onlineSavingsMsg'
Learn More

Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted, Thermo Scientific Chemicals

Catalog Number p-7052366
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
7783-90-6
AgCl
143.32
MFCD00003399
HKZLPVFGJNLROG-UHFFFAOYSA-M
Silver chloride dispersion in water
silver(1+) chloride
[Cl-].[Ag+]
This item is not returnable. View return policy
7783-90-6
AgCl
143.32
MFCD00003399
HKZLPVFGJNLROG-UHFFFAOYSA-M
Silver chloride dispersion in water
silver(1+) chloride
[Cl-].[Ag+]

Used as pharmaceutical intermediate and in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Density 1.038 g/mL
UN Number UN3082
Quantity 100 g
Merck Index 14,8509
Solubility Information Dispersible in water. Very slightly soluble in cold water,soluble in solutions of alkali cyanides,thiosulfates,ammonium carbonates. Appreciably soluble in concentrated aqueous solutions of ammonium chloride,mercuric nitrate,and silver nitrate.
Formula Weight 143.32
Concentration or Composition (by Analyte or Components) 4.9% in H2O
Odor Odorless
Particle Size <200nm APS Number Weighted
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Silver chloride, Nanoparticle dispersion
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'