Learn More
Thermo Scientific™ SilTite™ Ferrule Kits
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Brand:  Thermo Scientific™ 290MT231
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10724556
  • 158.29 EUR / Pack of 10
Estimated Shipment: 30-05-2024
Description
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
Specifications
Specifications
SilTite Kits for Thermo Scientific DSQ GC-MS Interface
SilTite Metal
Two SilTite Nuts and Ten Ferrules
290 MT231
Ferrule Kit
0.53 mm