missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Versilic™ Silicone Stoppers, 40mmH
Brand: Saint-Gobain 26304902
Additional Details : Weight : 0.11000kg
Description
- Biologically neutral and non-toxic silicone caps for closing tubes and flasks
- Odorless, tasteless, non-adherent and resistant to aging
- Nontoxic, biologically neutral
- FDA/BGA approved
- For test tubes
- Easy to install
Specifications
FDA compliant food contact / BFR | |
2/Pk. | |
Yes | |
Silicone |
40mm | |
–60 to 200°C | |
49mm |