Learn More
Cytiva Silicone Tubing
140mm length
Brand: Cytiva 19-4692-01
Additional Details : Weight : 0.00300kg
Description
Silicone tubing for Peristaltic Pump P-1.Specifications
Peristaltic Pump p-1 | |
Tubing | |
140 mm | |
Silicone |