missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silica gel, preparative chromatography grade, spherical, 7.5 micron APS, 80 angstroms, 99.99+%

Brand:  Alfa Aesar™ 045815.06

Additional Details : CAS Number : 63231-67-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15444975

  • 46.00 EUR / 5g


Estimated Shipment date
from Supplier 24-06-2019
Add to basket

Specifications

Chemical Identifiers

63231-67-4
78.098
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
H2O3Si
MFCD00011232
14768
O[Si](=O)O

Specifications

Silica gel
63231-67-4
H2O3Si
7.5 Micron APS, 80Å
O[Si](=O)O
78.098
Spherical
≥99.99%
P.V. 1.00cc/g
5g
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
14768
650m2/g
Preparative Chromatography
Videos
SDS Documents