Learn More
Alfa Aesar™ Silica gel, HPLC grade, spherical, 3 micron APS, 70 angstroms, 99.99+%
Brand:  Alfa Aesar™ 045802.03
Additional Details : CAS Number : 63231-67-4 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15487075
  • 65.00 EUR / 1g
Estimated Shipment date
from Supplier 06-08-2018
Add to basket
Specifications
Chemical Identifiers
63231-67-4
78.098
IJKVHSBPTUYDLN-UHFFFAOYSA-N
H2O3Si
MFCD00011232
14768
Specifications
Silica gel
P.V. 0.95cc/g
HPLC Grade
Spherical
H2O3Si
500m2/g
O[Si](=O)O
78.098
1g
63231-67-4
3 Micron APS, 70Å
≥99.99%
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
14768