Learn More
Silica Gel 40 Angstrom Pore Size, 0.2-0.5mm, for Chromatography Column, Fisher Chemical™
Brand: Fisher Chemical S/0780/60
Additional Details : CAS Number : 7631-86-9 Weight : 1.00000kg
Specifications
Plastic Powder Jar | |
5.5 to 8 | |
1 kg |
Brand: Fisher Chemical S/0780/60
Additional Details : CAS Number : 7631-86-9 Weight : 1.00000kg
Plastic Powder Jar | |
5.5 to 8 | |
1 kg |