Learn More
BRAND™ SilberBrand™ ETERNA Type 2 Graduated Pipettes
AR-Glas™ graduated pipettes
Brand:  BRAND™ 27009
Additional Details : Weight : 0.21110kg
 View more versions of this product
Product Code. 15613957
  • 61.20 EUR / Pack of 12
Estimated Shipment: 13-06-2024
Description
  • DIN EN ISO 835
  • Calibrated to deliver (TD, Ex)
  • Specifications
    Specifications
    To Deliver
    B
    Upper end cotton plug
    Red
    360 mm
    DIN EN ISO 835
    0.05 mL
    12
    5 mL