Learn More
Epredia™ Signature Series™ Cyto-Stain™
Achieve results similar to conventional Papanicolaou stains with Epredia™ Signature Series Cyto-Stain™, a uniquely formulated cytoplasmic stain
Brand:  Epredia™ 7511
Additional Details : Weight : 0.40000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
 View more versions of this product
Product Code. 10465680
  • 50.00 EUR / Pack of 4
Estimated Shipment: 23-08-2024
Description
  • Use for all gynecological and nongynecological procedures
  • Reduces staining time
  • Cost-effective, as user performs only one counterstain
  • Cyto-Stain provides a full range of familiar cytoplasmic colors  and control over the staining intensities
  • Cyto-Stain G is a modification to produce greener cyanophilic hues in intermediate and basal cells
Specifications
Specifications
Cytostain
4 pt.