Learn More
Trajan™ Shimadzu Instrument Ferrules
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
113.00 EUR - 133.00 EUR
Product Code | Brand | For Use With (Equipment) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Material | Price | Quantity & Availability | ||||
15258913
|
Trajan™
073204 |
QP5000/5050 Standard MS Interface | SilTite Metal |
133.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
15208923
|
Trajan™
073227 |
SGE; GC14 A, GC17 A, GC2010 and GC2014 Detector/Injectors | SilTite Metal |
118.00 EUR
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
15278883
|
Trajan™
0726564 |
QP5000/5050 Standard MS Interface | 15% Graphite, 85% Vespel |
113.00 EUR
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Ferrule |