Learn More
Justrite Shelves for EN Cabinet 226-EN 2903
Additional shelf accessory for EN Cabinets 226-EN. Choice of installed or pull out shelves in stainless steel or epoxy coated finishing.
Brand:  Justrite JCB60IS000
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 17712378
  • 125.00 EUR / Each
Estimated Shipment: 20-06-2024
Description
  • Choice of stainless steel or epoxy coated shelves
  • Choice of installed or pull out shelves
Specifications
Specifications
Stainless Steel
Shelf
Steel
42 cm
Pull Out
49 cm
EN Cabinet 226-EN 2903