Learn More
SGE™ SilFlow™ GC 3 Port Splitter Kits
Highly efficient and reliable micro fluidic platform to improve GC connectivity.
Brand: SGE™ 123720
Additional Details : Weight : 0.01000kg
Description
- Kit includes a SilFlow microchannel device (MCD), SilTite FingerTite tool, mounting bracket, plus appropriate ferrules, nuts and blanking ferrules for set up
- 1.1mm OD tubing not included
Specifications
Port Splitter | |
Port B and C 0.25/0.32 mm | |
SGE Silflow Microchannel Device |
1.1 mm | |
SilFlow microchannel device (MCD), SilTite FingerTite tool, Mounting bracket, Appropriately sized ferrules, nuts and blanking ferrules for set up |