Learn More
Trajan™ SilTite™ GC Capillary Column Connectors: μ Unions
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand: Trajan™ 073560
Additional Details : Weight : 0.20000kg
Description
- Low thermal mass
- Zero Dead Volume
- Inert
- No-need to retighten after installation
- Connect Pre-column or GC x GC
Specifications
Connector | |
0.1-0.25 mm and 0.1-0.25 mm | |
2 unions, 5 double taper ferrules and tools | |
GC Capillary Column |