Learn More
Trajan™ Hexnut Fittings
HexNut fittings provide ideal connections in HPLC systems
Brand:  Trajan™ 1021011
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15259103
  • 213.00 EUR / Pack of 10
Estimated Shipment: 29-05-2024
Description
  • Stainless steel 10-32 thread
  • Inert contact surface makes it biocompatible
  • Ideal for use with corrosive solvents
Specifications
Specifications
10 hexnuts and 10 ferrules
10-32
Stainless Steel
5000 psi