Learn More
Trajan™ Hexnut Fittings
HexNut fittings provide ideal connections in HPLC systems
47.80 EUR - 213.00 EUR
Specifications
Thread Size 10-32
Material Stainless Steel
Pressure 5000 psi
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
15249103
View Documents Promotion Details
Trajan™
1021003
2 hexnuts and 2 ferrules
47.80 EUR
Pack of 2
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
15259103
View Documents Promotion Details
Trajan™
1021011
10 hexnuts and 10 ferrules
213.00 EUR
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
Description
  • Stainless steel 10-32 thread
  • Inert contact surface makes it biocompatible
  • Ideal for use with corrosive solvents
Specifications
Specifications
10-32
Stainless Steel
5000 psi