Learn More
Trajan™ BPX Dioxin Capillary Columns
SGE BPX Dioxin GC Capillary Columns arespecifically designed for analysis or dioxins and related compounds
Brand: Trajan™ 054235
Additional Details : Weight : 0.01000kg
Description
- High temperature, low bleed column
- Non-polar
- Inert
Specifications
0.25 mm | |
0.25 μm | |
60 m |