Learn More
Robu™ Series 15 Filter Discs
Robu™ Series 15 Filter Discs feature a unique porous structure, which make them flexible and incredibly versatile for a variety of applications.
Brand:  Robu™ 15102
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12060402
  • 39.85 EUR / Pack of 10
Estimated Shipment: 26-06-2024
Description
  • Surfaces untreated
  • Ground cylindrical
  • The faces are press molded and may show thickness deviations
Specifications
Specifications
Untreated surface
40 to 100 μm
2.8 ± 0.7 mm
10 mm
Glass
Cylindrical
Filter Disc