Learn More
Robu™ Series 15 Filter Discs
Robu™ Series 15 Filter Discs feature a unique porous structure, which make them flexible and incredibly versatile for a variety of applications.
Brand:  Robu™ 15303
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12050422
  • 46.40 EUR / Pack of 10
Estimated Shipment: 26-06-2024
Description
  • Surfaces untreated
  • Ground cylindrical
  • The faces are press molded and may show thickness deviations
Specifications
Specifications
Untreated surface
16 to 40 μm
3.5 ± 0.8 mm
30 mm
Glass
Cylindrical
Filter Disc