Learn More
Fisherbrand™ Septum for 13-425 screw cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand:  Fisherbrand™
336.30 EUR valid until 2024-12-31
Use promo code "23262" to get your promotional price.
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 11844950
  • 354.00 EUR / Pack of 1000
Estimated Shipment: 24-05-2024
Description
  • 12mm Septa
Specifications
Specifications
Septa
12 mm
1000
Red, White
PTFE, Silicone
1 mm