Learn More
Hamilton™ Septa, Low Bleed High Temp
151.00 EUR
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10186262
|
Hamilton™
75835 |
11 mm |
151.00 EUR
Pack of 50 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
10024202
|
Hamilton™
75834 |
9 mm |
151.00 EUR
Pack of 50 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||