Learn More
Sartorius Vivapure™ AdenoPACK 500 Kit
Pump driven Kit for larger volumes
Brand:  Sartorius VS-AVPQ501
Additional Details : Weight : 0.01000kg
Product Code. 11740855
  • 1995.00 EUR / Each
Estimated Shipment: 13-06-2024
Description
Vivapure™ AdenoPACK 500 is the direct upscale kit to the AdenoPACK 100, for adenovirus purification. In only 2 hours up to 3 × 1013 adenovirus particles are purified and concentrated from 500 ml cell culture. This completely ready-to-use kit is conveniently operated by a laboratory pump, offering optimal flow control and minimal hands-on time. This easy to use product replaces lengthy and inefficient cesium chloride density gradient methods.
  • Pump driven kit
  • Easy and controlled purification of Adenovirus from large cell culture volumes
  • Virus purification based on membrane adsorber technology
  • High-speed process completed in about 2 hours
  • Convenient, easy to use alternative to CsCl density gradients
  • Able to process 10 to 25 plates (up to 500 ml)
  • Flexible, most cost-effective kit in the market for purifying 1-3 × 1013 viral particles
Specifications
Specifications
<0.025 EU/mL
500 mL of Adenovirus supernatants
-20°C