Learn More
Sartorius Vivaflow™ Models Accessories: Flow Restrictors
Product Code. 11730915 Shop All Sartorius Products
Click to view available options
For Use With (Equipment):
Vivaflow 50 and 200
Unit Size:
Pack of 6
Product Code. 11730915
Brand: Sartorius VFA035
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Flow Restrictors for Vivaflow™ 50 and 200.
  • Manage and maintain flow
TRUSTED_SUSTAINABILITY
Specifications
Color Pink
Filter Type Ultrafiltration
For Use With (Equipment) Vivaflow 50 and 200
Material PVC
Pore Size 0.6 mm
Sterility Non-sterile
Quantity 6 Pk.
Type Flow Restrictor