missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Quantitative Grade 389 F Filter Papers
Brand: Sartorius™ FT-3-112-150
Additional Details : Weight : 0.01000kg
Description
- Ash-free (ash content ≤ 0.01 % according to DIN 540370)
- Wet-strengthened
- Color-coded box for easy selection
Specifications
100 per pack | |
Disc | |
Yellow |
389 | |
150mm | |
0.19mm |