Learn More
Sartorius Arium™ Bagtank Rollers
Arium™ Bagtank Rollers enable Arium™ Bagtanks to be moved quickly and conveniently to wherever it is needed. Arium™ Bagtank Rollers are an optional accessory designed for use with Arium™ Bagtank 50 and Arium™ Bagtank 100 purified water storage units.
Brand: Sartorius H2O-ATR
Additional Details : Weight : 10.00000kg
Description
The closed Arium™ Bagtank system reliably protects purified water from secondary contamination to ensure consistently high water quality. Unlike a standard tank system, the bag can be replaced in less than 5 minutes and eliminates time-consuming cleaning procedures with hazardous chemicals.
- Optional rollers makes bagtanks extremely portable and flexible
- Used with the Arium™ Bagtank 50 or Bagtank 100
- Compatible with Arium™ Comfort and Arium™ Advance Water Purification Systems
Specifications
Arium Bagtank 50 and Bagtank 100 Ultrapure Water Storage System | |
Rollers | |
Storage of ultra-purified water | |
4 Rollers and Fittings |