Learn More
Sartorius Qualitative Grade 293 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand:  Sartorius FT-3-211-055
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11711195
  • 11.30 EUR / Pack of 100
Estimated Shipment: 01-07-2024
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
Disc
300 sec.
100/Pk.
55 mm
1 to 2 μm
293
0.15 mm