Learn More
Sartorius Qualitative Grade 293 Filter Papers
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Brand:  Sartorius FT-3-211-110
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 10322072
  • 16.10 EUR / Pack of 100
In Stock
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
Specifications
Specifications
Disc
300 sec.
100/Pk.
110 mm
1 to 2 μm
293
0.15 mm