Learn More
S Murray™ SAMCO™ Dissecting Scissors
Use these scissors for a variety of applications. S Murray™ SAMCO™ Dissecting Scissors feature fine points, open shanks and stainless steel construction.
4.15 EUR - 5.50 EUR
Product Code | Brand | Blade Style | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Blade Style | Length (Metric) | Price | Quantity & Availability | ||||
12952055
|
S Murray™
E103/02 |
Curved | 125 mm |
4.40 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
12338099
|
S Murray™
E101/02 |
Straight | 125 mm |
4.65 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
12328099
|
S Murray™
E101/01 |
Straight | 112 mm |
4.15 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
12902055
|
S Murray™
E101/03 |
Straight | 150 mm |
5.50 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
Description
- Stainless Steel dissecting scissors
Specifications
Dissecting Scissors | |
Stainless Steel |