Learn More
Thermo Scientific™ Saline 0.45% Solution Tubes
Reduce sample preparation time for automated identification and susceptibility systems using convenient, pre-filled and ready-to-use 0.45% saline tubes.
585.00 EUR
Specifications
Color | Clear liquid colorless |
---|---|
Content And Storage | 2 to 25°C |
Description | Saline 0.45% Solution Tubes |
Format | Tube |
Manufacturing Origin | United States |
Product Code | Brand | Description | Quantity | Fill Volume | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Fill Volume | Price | Quantity & Availability | |||
13205549
|
Thermo Scientific™
R064462 |
Saline 0.45% Solution Tubes | 100/Pk. | 1.8 mL |
585.00 EUR
Pack of 100 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
Description
Save time when preparing your samples for automated identification and susceptibility systems using Thermo Scientific™ Saline 0.45% Solution Tubes.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes.- Easily adjust inoculum density with a clear tube
- Enhance sample security while promoting safe handling with a reliable screw cap on each tube
- Achieve versatility and flexibility with fill volumes in 1.8mL and 3.0mL
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Clear liquid colorless | |
Saline 0.45% Solution Tubes | |
United States | |
100/Pk. |
2 to 25°C | |
Tube | |
Buffer Solution |