Learn More
Justrite™ Safety Cabinet Standard
Brand:  Justrite™ 8945021
Additional Details : Weight : 136.53130kg
Product Code. 11985426
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
2 door, manual, 2 adj shelves not for phenol, nitric /sulphuric acid, steel, double wall dual vents, earth wire connections, leakproof sills, levelling feet blue 1,651mm x 1,092mm x 457mm (w x h x d) 170L