missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-Allylglycine, 95%, Thermo Scientific Chemicals

Catalog Number p-3776595
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
90600-20-7
C10H17NO4
215.249
MFCD01320851
BUPDPLXLAKNJMI-ZETCQYMHSA-N
s-2-tert-butoxycarbonyl amino pent-4-enoic acid, s-n-boc-allylglycine, boc-l-allylglycine, boc-alpha-allyl-l-gly, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, s-2-tert-butoxycarbonylamino pent-4-enoic acid, 2s-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-l-alpha-allyl-gly, boc--allyl-l-gly, rarechem bk pt 0250
2734487
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
CC(C)(C)OC(=O)NC(CC=C)C(=O)O
This item is not returnable. View return policy
90600-20-7
C10H17NO4
215.249
MFCD01320851
BUPDPLXLAKNJMI-ZETCQYMHSA-N
s-2-tert-butoxycarbonyl amino pent-4-enoic acid, s-n-boc-allylglycine, boc-l-allylglycine, boc-alpha-allyl-l-gly, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, s-2-tert-butoxycarbonylamino pent-4-enoic acid, 2s-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-l-alpha-allyl-gly, boc--allyl-l-gly, rarechem bk pt 0250
2734487
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
CC(C)(C)OC(=O)NC(CC=C)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Assay Percent Range 95%
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation + 10.50
Formula Weight 215.25
Percent Purity 95%
Specific Rotation Condition +10.50° (20°C c=1,methanol)
Quantity 500 mg
Physical Form Viscous Liquid
Chemical Name or Material (S)-N-BOC-Allylglycine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'