missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%, Thermo Scientific Chemicals

Catalog Number p-3830592
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
17553-86-5
C10H12O2
164.2
MFCD00012272
FNYAZSZTENLTRT-JTQLQIEISA-N
7as-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione, s-+-2,3,7,7a-tetrahydro-7a-methyl-1h-indene-1,5 6h-dione, 1h-indene-1,5 6h-dione, 2,3,7,7a-tetrahydro-7a-methyl-, 7as, hajos dione, s-hajos dione, s-hajos ketone, s-hajos-wiechert ketone, +-hajos-parrish ketone, +-hajos-parrish diketone, 7as-7a-methyl-2,3,7,7a-tetrahydro-1h-indene-1,5 6h-dione
736943
(7aS)-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione
CC12CCC(=O)C=C1CCC2=O
This item is not returnable. View return policy
17553-86-5
C10H12O2
164.2
MFCD00012272
FNYAZSZTENLTRT-JTQLQIEISA-N
7as-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione, s-+-2,3,7,7a-tetrahydro-7a-methyl-1h-indene-1,5 6h-dione, 1h-indene-1,5 6h-dione, 2,3,7,7a-tetrahydro-7a-methyl-, 7as, hajos dione, s-hajos dione, s-hajos ketone, s-hajos-wiechert ketone, +-hajos-parrish ketone, +-hajos-parrish diketone, 7as-7a-methyl-2,3,7,7a-tetrahydro-1h-indene-1,5 6h-dione
736943
(7aS)-7a-methyl-2,3,6,7-tetrahydroindene-1,5-dione
CC12CCC(=O)C=C1CCC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Melting Point 55.0°C to 64.0°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition +331° (20°C c=1,toluene)
Specific Rotation + 331.00
Formula Weight 164.2
Percent Purity 90%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material (S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'