missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1,4-Diazabicyclo[4.3.0]nonane, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Quantity | 250 mg |
UN Number | UN3267 |
Solubility Information | Slightly soluble in water. |
IUPAC Name | (8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine |
Formula Weight | 126.2 |
Percent Purity | ≥98% |
Chemical Name or Material | (S)-1,4-Diazabicyclo[4.3.0]nonane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'