Learn More
Fisherbrand™ Round Gel Pipet Tips
Round gel loading tip measures 68.6mm long and fits most 20, 50, 100 and 200μL pipettors. Fisherbrand™ Round Gel Pipet Tips are to be used with agarose or acrylamide gels.
Brand:  Fisherbrand™
Additional Details : Weight : 0.05000kg
Product Code. 11572343
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Natural color
  • 200μL capacity
  • Provided in racks
  • Sterilized
Specifications
Specifications
1 to 200 μL
Gel Loading