Learn More
Lenz Laborglasinstrumente™ DURAN™ Round Bottom Flasks
Can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ DURAN™ Round Bottom Flasks are manufactured from durable DURAN glass.
Brand: Lenz Laborglasinstrumente™ B0418061
Additional Details : Weight : 0.05220kg
Description
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifications
100 mL | |
Borosilicate Glass, Duran Glass | |
NS 29 | |
Round |